Assay Details
| Gene Symbol: Atp6v1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007509.3 | NP_031535.2 | 5 - 6 | 518 | 72 | |
| GenBank mRNA | AK146499.1 | - | 5 - 6 | 483 | 72 | |
| AK151200.1 | - | 5 - 6 | 519 | 72 | ||
| AK151322.1 | - | 5 - 6 | 491 | 72 | ||
| AK151586.1 | - | 5 - 6 | 491 | 72 | ||
| AK152718.1 | - | 5 - 6 | 491 | 72 | ||
| AK152766.1 | - | 5 - 6 | 491 | 72 | ||
| AK153341.1 | - | 5 - 6 | 482 | 72 | ||
| AK159133.1 | - | 5 - 6 | 489 | 72 | ||
| AK159153.1 | - | 5 - 6 | 489 | 72 | ||
| AK159586.1 | - | 5 - 6 | 486 | 72 | ||
| AK159701.1 | - | 5 - 6 | 490 | 72 | ||
| AK159986.1 | - | 5 - 6 | 490 | 72 | ||
| AK160080.1 | - | 5 - 6 | 489 | 72 | ||
| AK160854.1 | - | 5 - 6 | 484 | 72 | ||
| AK166669.1 | - | 5 - 6 | 458 | 72 | ||
| AK168852.1 | - | 5 - 6 | 482 | 72 | ||
| AK169155.1 | - | 5 - 6 | 497 | 72 | ||
| AK169270.1 | - | 5 - 6 | 494 | 72 | ||
| AK169420.1 | - | 5 - 6 | 486 | 72 | ||
| AK170110.1 | - | 5 - 6 | 514 | 72 | ||
| AK182828.1 | - | 5 - 6 | 373 | 72 | ||
| AK185732.1 | - | 5 - 6 | 343 | 72 | ||
| AK187147.1 | - | 5 - 6 | 361 | 72 | ||
| AK191995.1 | - | 5 - 6 | 341 | 72 | ||
| AK200912.1 | - | 4 - 5 | 332 | 72 | ||
| AK204303.1 | - | 4 - 5 | 335 | 72 | ||
| AK207516.1 | - | 4 - 5 | 315 | 72 | ||
| AK208817.1 | - | 4 - 5 | 321 | 72 | ||
| AK209216.1 | - | 5 - 6 | 353 | 72 | ||
| AK209288.1 | - | 5 - 6 | 352 | 72 | ||
| AK210876.1 | - | 2 - 3 | 133 | 72 | ||
| AK214894.1 | - | 5 - 6 | 349 | 72 | ||
| AK215413.1 | - | 4 - 5 | 317 | 72 | ||
| AK216107.1 | - | 4 - 5 | 315 | 72 | ||
| AK216725.1 | - | 5 - 6 | 353 | 72 | ||
| AK220015.1 | - | 5 - 6 | 355 | 72 | ||
| BC012497.1 | - | 5 - 6 | 483 | 4500843 | 72 | |
| BC046302.1 | - | 5 - 6 | 473 | 6492612 | 72 | |
| BC085300.1 | - | 5 - 6 | 465 | 6305977 | 72 | |
| Y12634.1 | - | 5 - 6 | 475 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b2
Genomic Map