Assay Details
| Gene Symbol: Atp6v1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007509.3 | NP_031535.2 | 7 - 8 | 757 | 107 | |
| GenBank mRNA | AK146499.1 | - | 7 - 8 | 722 | 107 | |
| AK151200.1 | - | 7 - 8 | 758 | 107 | ||
| AK151322.1 | - | 7 - 8 | 730 | 107 | ||
| AK151586.1 | - | 7 - 8 | 730 | 107 | ||
| AK152718.1 | - | 7 - 8 | 730 | 107 | ||
| AK152766.1 | - | 7 - 8 | 730 | 107 | ||
| AK153341.1 | - | 7 - 8 | 721 | 107 | ||
| AK159133.1 | - | 7 - 8 | 728 | 107 | ||
| AK159153.1 | - | 7 - 8 | 728 | 107 | ||
| AK159586.1 | - | 7 - 8 | 725 | 107 | ||
| AK159701.1 | - | 7 - 8 | 729 | 107 | ||
| AK159986.1 | - | 7 - 8 | 729 | 107 | ||
| AK160080.1 | - | 7 - 8 | 728 | 107 | ||
| AK160854.1 | - | 7 - 8 | 723 | 107 | ||
| AK166669.1 | - | 7 - 8 | 697 | 107 | ||
| AK168852.1 | - | 7 - 8 | 721 | 107 | ||
| AK169155.1 | - | 7 - 8 | 736 | 107 | ||
| AK169270.1 | - | 7 - 8 | 733 | 107 | ||
| AK169420.1 | - | 7 - 8 | 725 | 107 | ||
| AK210829.1 | - | 2 - 3 | 107 | |||
| BC012497.1 | - | 7 - 8 | 722 | 4500843 | 107 | |
| BC046302.1 | - | 7 - 8 | 712 | 6492612 | 107 | |
| BC085300.1 | - | 7 - 8 | 704 | 6305977 | 107 | |
| Y12634.1 | - | 7 - 8 | 714 | 107 | ||
More Information
Set Membership: |
|

Atp6v1b2
Genomic Map