Assay Details
| Gene Symbol: Bmpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277216.1 | NP_001264145.1 | 3 - 4 | 308 | 84 | |
| NM_001277217.1 | NP_001264146.1 | 3 - 4 | 334 | 84 | ||
| NM_001277218.1 | NP_001264147.1 | 2 - 3 | 347 | 84 | ||
| NM_007560.4 | NP_031586.1 | 2 - 3 | 501 | 84 | ||
| GenBank mRNA | AK086130.1 | - | 3 - 4 | 294 | 84 | |
| AK162844.1 | - | 2 - 3 | 182 | 84 | ||
| BC065106.1 | - | 2 - 3 | 282 | 30357130 | 84 | |
| BC065143.1 | - | 2 - 3 | 488 | 30629345 | 84 | |
| BC138638.1 | - | 2 - 3 | 228 | 8861658 | 84 | |
| BC138640.1 | - | 2 - 3 | 228 | 8861660 | 84 | |
| CV557650.1 | - | 2 - 3 | 347 | 84 | ||
| Z23143.1 | - | 3 - 4 | 326 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmpr1b
Genomic Map