Assay Details
| Gene Symbol: Btc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007568.5 | NP_031594.1 | 4 - 5 | 698 | 105 | |
| GenBank mRNA | AK050206.1 | - | 4 - 5 | 514 | 105 | |
| AK076272.1 | - | 4 - 5 | 618 | 105 | ||
| BC119152.1 | - | 4 - 5 | 564 | 8733901 | 105 | |
| BC119154.1 | - | 4 - 5 | 564 | 8733903 | 105 | |
| BC144906.1 | - | 3 - 4 | 446 | 105 | ||
| L08394.1 | - | 4 - 5 | 537 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Btc
Genomic Map