Assay Details
| Gene Symbol: Camk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174054.1 | NP_001167525.1 | 15 - 16 | 1301 | 75 | |
| NM_007595.5 | NP_031621.3 | 15 - 16 | 1340 | 75 | ||
| GenBank mRNA | AK046454.1 | - | 15 - 16 | 1341 | 75 | |
| AK163698.1 | - | 15 - 16 | 1157 | 75 | ||
| BC024803.1 | - | 15 - 16 | 1117 | 75 | ||
| X63615.1 | - | 15 - 16 | 1155 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk2b
Genomic Map