Assay Details
| Gene Symbol: Camk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174053.1 | NP_001167524.1 | 17 - 18 | 1522 | 107 | |
| NM_001174054.1 | NP_001167525.1 | 18 - 19 | 1474 | 107 | ||
| NM_007595.5 | NP_031621.3 | 18 - 19 | 1513 | 107 | ||
| GenBank mRNA | AK046454.1 | - | 18 - 19 | 1514 | 107 | |
| AK158794.1 | - | 13 - 14 | 1157 | 107 | ||
| AK163698.1 | - | 18 - 19 | 1330 | 107 | ||
| BC024803.1 | - | 18 - 19 | 1290 | 107 | ||
| BC080273.1 | - | 17 - 18 | 1324 | 6851995 | 107 | |
| X63615.1 | - | 18 - 19 | 1328 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk2b
Genomic Map