Assay Details
| Gene Symbol: Cidea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007702.2 | NP_031728.2 | 2 - 3 | 421 | 62 | |
| GenBank mRNA | AF041376.1 | - | 2 - 3 | 395 | 62 | |
| BB614324.1 | - | 2 - 3 | 196 | 62 | ||
| BC096649.1 | - | 2 - 3 | 257 | 1397772 | 62 | |
| BQ554477.1 | - | 2 - 3 | 430 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cidea
Genomic Map