Assay Details
| Gene Symbol: Ccr1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007718.3 | NP_031744.3 | 1 | 952 | 65 | |
| GenBank mRNA | BC127143.1 | - | 1 | 952 | 40051964 | 65 |
| BC128466.1 | - | 1 | 952 | 40051965 | 65 | |
| CF551376.1 | - | 1 | 224 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chemotaxis
inflammatory response
immune response
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of monocyte chemotaxis
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
C-C chemokine receptor activity
chemotaxis
inflammatory response
immune response
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of monocyte chemotaxis
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
C-C chemokine receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004786 | BC127143.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004786 | BC128466.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004786 | NM_007718.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004786 | CF551376.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159463 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004786 | BC127143.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004786 | BC128466.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004786 | NM_007718.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004786 | CF551376.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214122 | BC127143.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214122 | BC128466.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214122 | NM_007718.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26897739 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159463 | - | - |
| Whole Mouse Genome G4122A | A_51_P214122 | BC127143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214122 | BC128466.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214122 | NM_007718.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117890 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159463 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07579775 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07579763 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07579755 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08044590 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07579757 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07579769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07550910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08044589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07579775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07788048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07550913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07579763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07550911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897743 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10598011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598012 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422349_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598010 | - | - |
| Mouse Genome 430 2.0 Array | 1422349_at | - | - |
| Mouse Genome 430 2.0 Array | 1422349_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12770_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12770_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900027982.mm.1 | NM_007718.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027982.mm.1 | NM_007718.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027982.mm.1 | TC0900003314.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422349_at | - | - |
| Mouse Expression Set 430 | 1422349_PM_at | - | - |
| Murine Genome U74v2 Set | 101733_PM_at | - | - |
| Murine Genome U74v2 Set | 101733_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422349_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422349_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422349_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422349_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 359588 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2450504 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002450504 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2625504 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2625504 | - | - |
| mouse-6 v1.1 (Illumina) | scl6336.1.1_233-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2450504 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl6336.1.1_233-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2450504 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2625504 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2625504 | - | - |

Ccr1l1
Genomic Map