Assay Details
| Gene Symbol: Dffb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007859.4 | NP_031885.3 | 5 - 6 | 782 | 85 | |
| GenBank mRNA | AB009377.1 | - | 5 - 6 | 690 | 85 | |
| AK077619.1 | - | 5 - 6 | 828 | 85 | ||
| BC053052.1 | - | 5 - 6 | 712 | 6403143 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dffb
Genomic Map