Assay Details
| Gene Symbol: Dnase1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007870.3 | NP_031896.3 | 5 - 6 | 760 | 62 | |
| GenBank mRNA | AF047355.1 | - | 5 - 6 | 735 | 62 | |
| AK156109.1 | - | 593 | 62 | |||
| AK171843.1 | - | 5 - 6 | 593 | 62 | ||
| AK172254.1 | - | 5 - 6 | 1153 | 62 | ||
| BC012671.1 | - | 5 - 6 | 713 | 4160709 | 62 | |
| U76110.1 | - | 5 - 6 | 569 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnase1l3
Genomic Map