Assay Details
| Gene Symbol: Dnmt3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271753.1 | NP_001258682.1 | 7 - 8 | 1112 | 61 | |
| NM_007872.4 | NP_031898.1 | 7 - 8 | 1158 | 61 | ||
| NM_153743.4 | NP_714965.1 | 2 - 3 | 336 | 61 | ||
| GenBank mRNA | AF068625.2 | - | 7 - 8 | 1062 | 61 | |
| AF480164.1 | - | 2 - 3 | 336 | 61 | ||
| AK077792.1 | - | 2 - 3 | 301 | 61 | ||
| AK090132.1 | - | 7 - 8 | 1089 | 61 | ||
| AK136170.1 | - | 2 - 3 | 258 | 61 | ||
| AK147263.1 | - | 7 - 8 | 1112 | 61 | ||
| AK147627.1 | - | 7 - 8 | 1112 | 61 | ||
| AK147642.1 | - | 7 - 8 | 1112 | 61 | ||
| AK147676.1 | - | 7 - 8 | 1087 | 61 | ||
| AK157792.1 | - | 2 - 3 | 307 | 61 | ||
| BC007466.1 | - | 7 - 8 | 1090 | 3492853 | 61 | |
| BE855330.1 | - | 2 - 3 | 336 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnmt3a
Genomic Map