Assay Details
| Gene Symbol: Slc26a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007885.2 | NP_031911.1 | 1 - 2 | 206 | 119 | |
| GenBank mRNA | AK052942.1 | - | 1 - 2 | 207 | 119 | |
| AK163974.1 | - | 1 - 2 | 97 | 119 | ||
| D42049.1 | - | 1 - 2 | 91 | 118 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK052942.1,AK163974.1,D42049.1,NM_007885.2 transcripts.
Set Membership: |
|

Slc26a2
Genomic Map