Assay Details
| Gene Symbol: Slc26a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007885.2 | NP_031911.1 | 2 - 3 | 335 | 107 | |
| GenBank mRNA | AK007212.1 | - | 2 - 3 | 401 | 107 | |
| AK052942.1 | - | 2 - 3 | 336 | 107 | ||
| AK163974.1 | - | 2 - 3 | 226 | 107 | ||
| BC028345.1 | - | 2 - 3 | 212 | 4014785 | 107 | |
| D42049.1 | - | 2 - 3 | 220 | 107 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK007212.1,AK052942.1,AK163974.1,BC028345.1,D42049.1,NM_007885.2 transcripts.
Set Membership: |
|

Slc26a2
Genomic Map