Assay Details
| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113389.1 | NP_001106860.1 | 5 - 6 | 656 | 78 | |
| NM_007890.2 | NP_031916.1 | 5 - 6 | 915 | 78 | ||
| GenBank mRNA | BC129888.1 | - | 4 - 5 | 518 | 40111469 | 78 |
| BC129889.1 | - | 4 - 5 | 518 | 40111470 | 78 | |
| BC138716.1 | - | 4 - 5 | 545 | 8861738 | 78 | |
| BC145405.1 | - | 4 - 5 | 518 | 9053963 | 78 | |
| U58497.1 | - | 4 - 5 | 545 | 78 | ||
More Information
Set Membership: |
|

Dyrk1a
Genomic Map