Assay Details
| Gene Symbol: Eef2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267710.1 | NP_001254639.1 | 15 - 16 | 2025 | 81 | |
| NM_001267711.1 | NP_001254640.1 | 15 - 16 | 2020 | 81 | ||
| NM_007908.4 | NP_031934.1 | 15 - 16 | 2109 | 81 | ||
| GenBank mRNA | AK028524.1 | - | 15 - 16 | 2086 | 81 | |
| AK080915.1 | - | 15 - 16 | 2026 | 81 | ||
| AK135891.1 | - | 13 - 14 | 1451 | 81 | ||
| AK136015.1 | - | 14 - 15 | 1751 | 81 | ||
| AK195875.1 | - | 1 - 2 | 159 | 81 | ||
| BC003433.1 | - | 15 - 16 | 2057 | 2646503 | 81 | |
| BC055361.1 | - | 15 - 16 | 2020 | 30046537 | 81 | |
| U93848.1 | - | 14 - 15 | 1763 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eef2k
Genomic Map