| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900009719.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009719.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009719.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009719.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009719.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009717.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009717.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009717.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009717.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009717.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009713.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009713.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009713.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009713.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009713.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009720.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009720.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009720.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009720.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009720.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009710.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009710.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009710.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009710.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009710.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009718.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009718.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009718.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009718.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009718.mm.1 |
TC1900001039.mm.1 |
MTA TC |