Assay Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007937.3 | NP_031963.2 | 7 - 8 | 2248 | 69 | |
| GenBank mRNA | BU707304.1 | - | 3 - 4 | 268 | 69 | |
| U07357.1 | - | 7 - 8 | 1788 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha5
Genomic Map