Assay Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007937.3 | NP_031963.2 | 8 - 9 | 2319 | 79 | |
| GenBank mRNA | AK089143.1 | - | 9 - 10 | 2604 | 79 | |
| BC057401.1 | - | 7 - 8 | 2228 | 5697758 | 79 | |
| BU707304.1 | - | 4 - 5 | 339 | 79 | ||
| U07357.1 | - | 8 - 9 | 1859 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha5
Genomic Map