Assay Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007937.3 | NP_031963.2 | 10 - 11 | 2630 | 90 | |
| GenBank mRNA | BC057401.1 | - | 9 - 10 | 2539 | 5697758 | 90 |
| U07357.1 | - | 10 - 11 | 2170 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha5
Genomic Map