Assay Details
| Gene Symbol: Epo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007942.2 | NP_031968.1 | 2 - 3 | 222 | 64 | |
| GenBank mRNA | BC119265.1 | - | 2 - 3 | 222 | 8734014 | 64 |
| BC119271.1 | - | 2 - 3 | 222 | 8734020 | 64 | |
| BC144883.1 | - | 2 - 3 | 222 | 9053431 | 64 | |
| BC144887.1 | - | 2 - 3 | 222 | 9053435 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Epo
Genomic Map