Assay Details
| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008072.2 | NP_032098.2 | 7 - 8 | 956 | 71 | |
| GenBank mRNA | BC116832.1 | - | 7 - 8 | 891 | 40126151 | 71 |
| BC116836.1 | - | 7 - 8 | 891 | 40126155 | 71 | |
| S42882.1 | - | 7 - 8 | 949 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabrd
Genomic Map