Assay Details
| Gene Symbol: Gcdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044744.1 | NP_001038209.2 | 5 - 6 | 544 | 65 | |
| NM_008097.2 | NP_032123.3 | 4 - 5 | 601 | 65 | ||
| GenBank mRNA | AK078866.1 | - | 2 - 3 | 154 | 65 | |
| AK165406.1 | - | 4 - 5 | 427 | 65 | ||
| AU080952.1 | - | 5 - 6 | 401 | 65 | ||
| BC061158.1 | - | 4 - 5 | 431 | 30314273 | 65 | |
| U18992.1 | - | 4 - 5 | 382 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcdh
Genomic Map