Assay Details
| Gene Symbol: Gcdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044744.1 | NP_001038209.2 | 7 - 8 | 844 | 93 | |
| NM_008097.2 | NP_032123.3 | 6 - 7 | 901 | 93 | ||
| GenBank mRNA | AK078866.1 | - | 4 - 5 | 454 | 93 | |
| AK165406.1 | - | 6 - 7 | 727 | 93 | ||
| BC061158.1 | - | 6 - 7 | 731 | 30314273 | 93 | |
| U18992.1 | - | 6 - 7 | 682 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcdh
Genomic Map