Assay Details
| Gene Symbol: Gcdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044744.1 | NP_001038209.2 | 11 - 12 | 1455 | 73 | |
| NM_008097.2 | NP_032123.3 | 10 - 11 | 1512 | 73 | ||
| GenBank mRNA | AK078866.1 | - | 8 - 9 | 1065 | 73 | |
| AK079500.1 | - | 4 - 5 | 563 | 73 | ||
| AK165406.1 | - | 10 - 11 | 1338 | 73 | ||
| BC061158.1 | - | 10 - 11 | 1342 | 30314273 | 73 | |
| U18992.1 | - | 10 - 11 | 1293 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
acyl-CoA metabolic process
metabolic process
fatty acid oxidation
fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
glutaryl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
flavin adenine dinucleotide binding
acyl-CoA metabolic process
metabolic process
fatty acid oxidation
fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
glutaryl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
flavin adenine dinucleotide binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986441 | NM_008097.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986441 | AK078866.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986441 | NM_001044744.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117136 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131049 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986441 | NM_008097.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986441 | AK078866.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986441 | NM_001044744.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260098 | U18992.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022545 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01784312 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131049 | - | - |
| Whole Mouse Genome G4122A | A_51_P260098 | U18992.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105353 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131049 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06742072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06742058 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06742054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07304640 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06742071 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07304635 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06742067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07304638 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06742077 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06742051 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06742073 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07304641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07010251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06742052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06742066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06742055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06742054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06850128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06742072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06742075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26022459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06742078 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10580242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580245 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580237 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580238 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580239 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580241 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580236 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448717_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10580233 | - | - |
| Mouse Genome 430 2.0 Array | 1448717_at | - | - |
| Mouse Genome 430 2.0 Array | 1448717_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 270076_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 270076_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800019826.mm.1 | NM_001044744.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019826.mm.1 | NM_001044744.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019826.mm.1 | NM_008097.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019826.mm.1 | NM_008097.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019826.mm.1 | TC0800002594.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800019825.mm.1 | NM_001044744.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019825.mm.1 | NM_001044744.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019825.mm.1 | NM_008097.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019825.mm.1 | NM_008097.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019825.mm.1 | TC0800002594.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448717_PM_at | - | - |
| Mouse Expression Set 430 | 1448717_at | - | - |
| Murine Genome U74v2 Set | 160194_PM_at | - | - |
| Murine Genome U74v2 Set | 162189_PM_r_at | - | - |
| Murine Genome U74v2 Set | 162189_r_at | - | - |
| Murine Genome U74v2 Set | 160194_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448717_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448717_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448717_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448717_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2190324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2190324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4280347 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002190324 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280347 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222337 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228907 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228907 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222337 | - | - |
| mouse-6 v1.1 (Illumina) | scl000730.1_110-S | - | - |
| mouse-6 v1.1 (Illumina) | scl34554.11.1_42-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280347 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2190324 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000730.1_110-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34554.11.1_42-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280347 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190324 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228907 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3136542 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1222337 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222337 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3136542 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228907 | - | - |

Gcdh
Genomic Map