Assay Details
| Gene Symbol: Gfra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF002701.1 | - | 1 - 2 | 41 | 70 | |
| AK138512.1 | - | 1 - 2 | 767 | 70 | ||
| AK149349.1 | - | 1 - 2 | 771 | 70 | ||
| AK154579.1 | - | 1 - 2 | 777 | 70 | ||
| BC138561.1 | - | 1 - 2 | 162 | 8861581 | 70 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK149349.1 transcripts.
Set Membership: |
|

Gfra2
Genomic Map