Assay Details
| Gene Symbol: Gja8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008123.3 | NP_032149.1 | 2 | 637 | 64 | |
| GenBank mRNA | BC120513.1 | - | 2 | 558 | 40129436 | 64 |
| BC125569.1 | - | 2 | 558 | 40130084 | 64 | |
| GU363513.1 | - | 2 | 516 | 64 | ||
| GU363514.1 | - | 2 | 606 | 64 | ||
| GU363515.1 | - | 1 | 823 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2160837 | GU363515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2160837 | GU363514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2160837 | NM_008123.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2160837 | BC120513.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2160837 | BC125569.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674318 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655506 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103879 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2160837 | GU363515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2160837 | GU363514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2160837 | NM_008123.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2160837 | BC120513.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2160837 | BC125569.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P308862 | GU363515.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P308862 | GU363514.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P308862 | NM_008123.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P308862 | BC120513.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P308862 | BC125569.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22351886 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04998950 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103879 | - | - |
| Whole Mouse Genome G4122A | A_51_P308862 | GU363515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P308862 | GU363514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P308862 | NM_008123.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P308862 | BC120513.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P308862 | BC125569.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114815 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103879 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02754305 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02754312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02754307 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02754313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22351841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03059793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02449328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02856731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02525927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03195294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02460914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02940893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02867883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02603855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02754310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02460913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02616826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02754308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02856732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03195295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02998378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02343065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22351909 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500432 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422274_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500429 | - | - |
| Mouse Genome 430 2.0 Array | 1422274_at | - | - |
| Mouse Genome 430 2.0 Array | 1422274_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14616_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14616_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300019751.mm.1 | NM_008123.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019751.mm.1 | TC0300002532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019754.mm.1 | NM_008123.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019754.mm.1 | TC0300002532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019752.mm.1 | NM_008123.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019752.mm.1 | TC0300002532.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422274_PM_at | - | - |
| Mouse Expression Set 430 | 1422274_at | - | - |
| Murine Genome U74v2 Set | 101286_at | - | - |
| Murine Genome U74v2 Set | 101286_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422274_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422274_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422274_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422274_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 702717 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7000619 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007000619 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2625168 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2625168 | - | - |
| mouse-6 v1.1 (Illumina) | scl21796.2_333-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7000619 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21796.2_333-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7000619 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2625168 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2625168 | - | - |

Gja8
Genomic Map