Assay Details
| Gene Symbol: Slc6a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008135.4 | NP_032161.2 | 9 - 10 | 1507 | 90 | |
| GenBank mRNA | AK178536.1 | - | 1 - 2 | 190 | 90 | |
| BC021828.1 | - | 9 - 10 | 1330 | 4194951 | 90 | |
| X67056.1 | - | 9 - 10 | 1440 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a9
Genomic Map