Assay Details
| Gene Symbol: Slc6a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008135.4 | NP_032161.2 | 10 - 11 | 1641 | 68 | |
| GenBank mRNA | AK014572.1 | - | 1596 | 68 | ||
| AK048168.1 | - | 1526 | 68 | |||
| BC021828.1 | - | 10 - 11 | 1464 | 4194951 | 68 | |
| X67056.1 | - | 10 - 11 | 1574 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a9
Genomic Map