Assay Details
| Gene Symbol: Gucy2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008192.3 | NP_032218.2 | 10 - 11 | 2207 | 140 | |
| GenBank mRNA | AK044282.1 | - | 10 - 11 | 2208 | 140 | |
| BC057954.1 | - | 10 - 11 | 2192 | 140 | ||
| BI738500.1 | - | 1 - 2 | 159 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2e
Genomic Map