Assay Details
| Gene Symbol: Hspe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008303.4 | NP_032329.1 | 1 - 2 | 262 | 97 | |
| GenBank mRNA | AK088121.1 | - | 1 - 2 | 93 | 97 | |
| AK133876.1 | - | 1 - 2 | 69 | 97 | ||
| AK151935.1 | - | 1 - 2 | 72 | 97 | ||
| AV570511.1 | - | 1 - 2 | 262 | 97 | ||
| BC099385.1 | - | 1 - 2 | 73 | 30848267 | 97 | |
| U09659.1 | - | 1 - 2 | 78 | 97 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK088121.1,AK133876.1,AK151935.1,BC099385.1,NM_008303.4,U09659.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20240346 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20240345 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20240347 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20240344 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089358 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124952 | AK088121.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710620 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139706 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157517 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124952 | AK088121.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953063 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089358 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436368 | U09659.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436368 | BC099385.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436368 | CJ238411.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436368 | NM_008303.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436368 | AK088121.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20240369 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157517 | - | - |
| Whole Mouse Genome G4122A | A_51_P436368 | U09659.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436368 | BC099385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436368 | CJ238411.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436368 | NM_008303.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436368 | AK088121.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124952 | AK088121.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122703 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124339 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104798 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157517 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00881081 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00881080 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20240344 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20240346 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20240345 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20240347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20240373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00881089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00881080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00453399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00871013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00248358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00599024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01100301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20240383 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10346307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455590 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346306 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346308 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450668_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422579_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10455588 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10346303 | - | - |
| Mouse Genome 430 2.0 Array | 1450668_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1450668_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422579_at | - | - |
| Mouse Genome 430 2.0 Array | 1422579_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15528_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15528_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100003103.mm.1 | NM_008303.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003103.mm.1 | NM_008303.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003103.mm.1 | TC0100000395.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100003107.mm.1 | NM_008303.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003107.mm.1 | NM_008303.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003107.mm.1 | TC0100000395.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100003105.mm.1 | NM_008303.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003105.mm.1 | NM_008303.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003105.mm.1 | TC0100000395.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100003101.mm.1 | NM_008303.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003101.mm.1 | NM_008303.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003101.mm.1 | TC0100000395.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100003108.mm.1 | NM_008303.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003108.mm.1 | NM_008303.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003108.mm.1 | TC0100000395.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450668_s_at | - | - |
| Mouse Expression Set 430 | 1450668_PM_s_at | - | - |
| Mouse Expression Set 430 | 1422579_PM_at | - | - |
| Mouse Expression Set 430 | 1422579_at | - | - |
| Murine Genome U74v2 Set | 92829_at | - | - |
| Murine Genome U74v2 Set | 92829_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450668_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450668_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422579_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422579_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450668_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422579_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450668_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422579_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 494056 | - | - |
| ABI Mouse Genome Survey Microarray | 903346 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3170438 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170438 | - | - |
| mouse-6 v1.1 (Illumina) | scl015528.3_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170438 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl015528.3_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170438 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2960308 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213364 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2960308 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl015528.3_20-S | - | - |

Hspe1
Genomic Map