Assay Details
| Gene Symbol: Htr1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008310.3 | NP_032336.1 | 2 - 3 | 268 | 121 | |
| GenBank mRNA | AK045364.1 | - | 2 - 3 | 337 | 121 | |
| BC120507.1 | - | 2 - 3 | 124 | 40129430 | 121 | |
| BC137604.1 | - | 2 - 3 | 124 | 8860623 | 120 | |
| Z14224.1 | - | 2 - 3 | 268 | 120 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK045364.1,BC120507.1,BC137604.1,NM_008310.3,Z14224.1 transcripts.
Set Membership: |
|

Htr1f
Genomic Map