Assay Details
| Gene Symbol: Htr2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008312.4 | NP_032338.3 | 4 - 5 | 1042 | 101 | |
| GenBank mRNA | AK034512.1 | - | 4 - 5 | 1044 | 101 | |
| AK079648.1 | - | 4 - 5 | 1044 | 101 | ||
| BC098327.1 | - | 5 - 6 | 1157 | 101 | ||
| BC141085.1 | - | 4 - 5 | 621 | 9055697 | 101 | |
| M63685.1 | - | 4 - 5 | 905 | 101 | ||
| X72230.1 | - | 4 - 5 | 906 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Htr2c
Genomic Map