Assay Details
| Gene Symbol: Il2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008368.4 | NP_032394.1 | 3 - 4 | 352 | 69 | |
| GenBank mRNA | AK017288.1 | - | 3 - 4 | 352 | 69 | |
| AK035289.1 | - | 3 - 4 | 305 | 69 | ||
| AK157572.1 | - | 3 - 4 | 353 | 69 | ||
| AK169481.1 | - | 3 - 4 | 333 | 69 | ||
| BC026869.1 | - | 3 - 4 | 325 | 4015306 | 69 | |
| M28052.1 | - | 3 - 4 | 309 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il2rb
Genomic Map