Assay Details
| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145802.1 | NP_001139274.1 | 10 - 11 | 1704 | 92 | |
| NM_001145803.1 | NP_001139275.1 | 11 - 12 | 1797 | 92 | ||
| NM_008547.2 | NP_032573.2 | 10 - 11 | 1692 | 92 | ||
| GenBank mRNA | BC050009.1 | - | 10 - 11 | 5567 | 92 | |
| X66983.1 | - | 10 - 11 | 1686 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mak
Genomic Map