Assay Details
| Gene Symbol: Mitf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113198.1 | NP_001106669.1 | 5 - 6 | 885 | 63 | |
| NM_001178049.1 | NP_001171520.1 | 5 - 6 | 772 | 63 | ||
| NM_008601.3 | NP_032627.1 | 4 - 5 | 576 | 63 | ||
| GenBank mRNA | AB221657.1 | - | 4 - 5 | 438 | 63 | |
| AK052895.1 | - | 5 - 6 | 744 | 63 | ||
| AK154189.1 | - | 5 - 6 | 886 | 63 | ||
| AK155350.1 | - | 5 - 6 | 886 | 63 | ||
| BC108976.2 | - | 4 - 5 | 540 | 40047440 | 63 | |
| BC108977.1 | - | 4 - 5 | 505 | 40047441 | 63 | |
| Z23066.1 | - | 4 - 5 | 567 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mitf
Genomic Map