Assay Details
| Gene Symbol: Ppp1r15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008654.2 | NP_032680.1 | 1 - 2 | 275 | 93 | |
| GenBank mRNA | AK079339.1 | - | 1 - 2 | 232 | 93 | |
| AK136336.1 | - | 1 - 2 | 232 | 93 | ||
| AK167317.1 | - | 1 - 2 | 231 | 93 | ||
| BC138485.1 | - | 1 - 2 | 140 | 8861505 | 93 | |
| BC138486.1 | - | 1 - 2 | 140 | 8861506 | 93 | |
| X51829.1 | - | 1 - 2 | 220 | 93 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK079339.1,AK167317.1,BC138485.1,BC138486.1,NM_008654.2,X51829.1 transcripts.
Set Membership: |
|

Ppp1r15a
Genomic Map