| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500009805.mm.1 |
NM_001039181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009805.mm.1 |
NM_001039181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009805.mm.1 |
NM_008728.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009805.mm.1 |
NM_008728.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009805.mm.1 |
TC1500001215.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500009804.mm.1 |
NM_001039181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009804.mm.1 |
NM_001039181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009804.mm.1 |
NM_001286395.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009804.mm.1 |
NM_008728.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009804.mm.1 |
NM_008728.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009804.mm.1 |
TC1500001215.mm.1 |
MTA TC |