Assay Details
| Gene Symbol: P2rx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008771.3 | NP_032797.3 | 3 - 4 | 526 | 71 | |
| GenBank mRNA | AK035304.1 | - | 3 - 4 | 527 | 71 | |
| BC015084.1 | - | 3 - 4 | 540 | 4189541 | 71 | |
| X84896.1 | - | 3 - 4 | 520 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P2rx1
Genomic Map