Assay Details
| Gene Symbol: P2ry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | L14751.1 | - | 1 - 2 | 267 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the L14751.1 transcripts.
Set Membership: |
|

P2ry2
Genomic Map