Assay Details
| Gene Symbol: Pip4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008845.4 | NP_032871.3 | 3 - 4 | 464 | 83 | |
| GenBank mRNA | AB009615.1 | - | 3 - 4 | 389 | 83 | |
| AK012196.1 | - | 3 - 4 | 467 | 83 | ||
| AK040558.1 | - | 3 - 4 | 354 | 83 | ||
| BC011097.1 | - | 3 - 4 | 411 | 3672732 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pip4k2a
Genomic Map