Assay Details
| Gene Symbol: Prkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008858.3 | NP_032884.2 | 7 - 8 | 1248 | 60 | |
| GenBank mRNA | Z34524.1 | - | 7 - 8 | 1123 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkd1| Gene Symbol: Prkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008858.3 | NP_032884.2 | 7 - 8 | 1248 | 60 | |
| GenBank mRNA | Z34524.1 | - | 7 - 8 | 1123 | 60 | |
Set Membership: |
|