Assay Details
| Gene Symbol: Serpine1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008871.2 | NP_032897.2 | 4 - 5 | 846 | 114 | |
| GenBank mRNA | BC054091.1 | - | 4 - 5 | 843 | 30021925 | 114 |
| GQ872458.1 | - | 3 - 4 | 701 | 114 | ||
| GQ872459.1 | - | 3 - 4 | 701 | 114 | ||
| M33960.1 | - | 4 - 5 | 832 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpine1
Genomic Map