Assay Details
| Gene Symbol: Por | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC003240.1 | - | 12 - 13 | 1492 | 3497749 | 60 |
| BC031463.1 | - | 12 - 13 | 1437 | 3497979 | 60 | |
| D17571.1 | - | 12 - 13 | 1436 | 60 | ||
More Information
Set Membership: |
|

Por
Genomic Map