Assay Details
| Gene Symbol: Map2k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008928.4 | NP_032954.1 | 1 - 2 | 216 | 103 | |
| GenBank mRNA | AK008141.1 | - | 1 - 2 | 219 | 103 | |
| AK011002.1 | - | 1 - 2 | 217 | 103 | ||
| AK053378.1 | - | 1 - 2 | 217 | 103 | ||
| AK150067.1 | - | 1 - 2 | 217 | 103 | ||
| AK166255.1 | - | 1 - 2 | 218 | 103 | ||
| AK166507.1 | - | 1 - 2 | 217 | 103 | ||
| AK167209.1 | - | 1 - 2 | 217 | 103 | ||
| AK167442.1 | - | 1 - 2 | 220 | 103 | ||
| AK168710.1 | - | 1 - 2 | 199 | 103 | ||
| BC007467.1 | - | 1 - 2 | 207 | 3497714 | 103 | |
| D87115.1 | - | 1 - 2 | 229 | 103 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008141.1,AK011002.1,AK150067.1,AK166255.1,AK166507.1,AK167209.1,AK167442.1,AK168710.1,BC007467.1,D87115.1,NM_008928.4 transcripts.
Set Membership: |
|

Map2k3
Genomic Map