Assay Details
| Gene Symbol: Map2k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008928.4 | NP_032954.1 | 2 - 3 | 278 | 78 | |
| GenBank mRNA | AK008141.1 | - | 2 - 3 | 281 | 78 | |
| AK011002.1 | - | 2 - 3 | 279 | 78 | ||
| AK053378.1 | - | 2 - 3 | 279 | 78 | ||
| AK150067.1 | - | 2 - 3 | 279 | 78 | ||
| AK159379.1 | - | 2 - 3 | 277 | 78 | ||
| AK166255.1 | - | 2 - 3 | 280 | 78 | ||
| AK166507.1 | - | 2 - 3 | 279 | 78 | ||
| AK167209.1 | - | 2 - 3 | 279 | 78 | ||
| AK167442.1 | - | 2 - 3 | 282 | 78 | ||
| AK168710.1 | - | 2 - 3 | 261 | 78 | ||
| BC007467.1 | - | 2 - 3 | 269 | 3497714 | 78 | |
| D87115.1 | - | 2 - 3 | 291 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k3
Genomic Map