Assay Details
| Gene Symbol: Map2k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008928.4 | NP_032954.1 | 4 - 5 | 444 | 65 | |
| GenBank mRNA | AK008141.1 | - | 4 - 5 | 447 | 65 | |
| AK011002.1 | - | 4 - 5 | 445 | 65 | ||
| AK053378.1 | - | 4 - 5 | 445 | 65 | ||
| AK150067.1 | - | 4 - 5 | 445 | 65 | ||
| AK159379.1 | - | 4 - 5 | 443 | 65 | ||
| AK166255.1 | - | 4 - 5 | 446 | 65 | ||
| AK166507.1 | - | 4 - 5 | 445 | 65 | ||
| AK167209.1 | - | 4 - 5 | 445 | 65 | ||
| AK167442.1 | - | 4 - 5 | 448 | 65 | ||
| AK168710.1 | - | 4 - 5 | 427 | 65 | ||
| BC007467.1 | - | 4 - 5 | 435 | 3497714 | 65 | |
| D87115.1 | - | 4 - 5 | 457 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k3
Genomic Map