Assay Details
| Gene Symbol: Map2k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008928.4 | NP_032954.1 | 6 - 7 | 686 | 88 | |
| GenBank mRNA | AK008141.1 | - | 6 - 7 | 689 | 88 | |
| AK011002.1 | - | 6 - 7 | 687 | 88 | ||
| AK053378.1 | - | 6 - 7 | 687 | 88 | ||
| AK150067.1 | - | 6 - 7 | 687 | 88 | ||
| AK159379.1 | - | 6 - 7 | 685 | 88 | ||
| AK166255.1 | - | 6 - 7 | 688 | 88 | ||
| AK166507.1 | - | 6 - 7 | 687 | 88 | ||
| AK167209.1 | - | 6 - 7 | 687 | 88 | ||
| AK167442.1 | - | 6 - 7 | 690 | 88 | ||
| AK168710.1 | - | 6 - 7 | 669 | 88 | ||
| D87115.1 | - | 6 - 7 | 699 | 88 | ||
| X93150.1 | - | 4 - 5 | 421 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k3
Genomic Map