Assay Details
| Gene Symbol: Ptger4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008965.2 | NP_032991.1 | 1 - 2 | 502 | 63 | |
| GenBank mRNA | AK145045.1 | - | 1 - 2 | 502 | 63 | |
| AK156428.1 | - | 1 - 2 | 504 | 63 | ||
| D13458.1 | - | 1 - 2 | 146 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK145045.1,D13458.1,NM_008965.2 transcripts.
Set Membership: |
|

Ptger4
Genomic Map