Assay Details
| Gene Symbol: Kif20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166406.1 | NP_001159878.1 | 11 - 12 | 1605 | 60 | |
| NM_001166407.1 | NP_001159879.1 | 11 - 12 | 1603 | 60 | ||
| NM_009004.4 | NP_033030.1 | 11 - 12 | 1499 | 60 | ||
| GenBank mRNA | AK051201.1 | - | 11 - 12 | 1417 | 60 | |
| AK051995.1 | - | 11 - 12 | 1454 | 60 | ||
| AK053280.1 | - | 11 - 12 | 1418 | 60 | ||
| AK083412.1 | - | 11 - 12 | 1459 | 60 | ||
| AK083697.1 | - | 11 - 12 | 1418 | 60 | ||
| AK084732.1 | - | 11 - 12 | 1441 | 60 | ||
| AK142764.1 | - | 11 - 12 | 1447 | 60 | ||
| AK144844.1 | - | 11 - 12 | 1417 | 60 | ||
| BC060608.1 | - | 11 - 12 | 1605 | 5720408 | 60 | |
| Y09632.1 | - | 11 - 12 | 1654 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif20a
Genomic Map