Assay Details
| Gene Symbol: Ralgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145834.1 | NP_001139306.1 | 9 - 10 | 1763 | 70 | |
| NM_001145835.1 | NP_001139307.1 | 9 - 10 | 1675 | 70 | ||
| NM_001145836.1 | NP_001139308.1 | 9 - 10 | 1639 | 70 | ||
| NM_009058.2 | NP_033084.2 | 9 - 10 | 1799 | 70 | ||
| GenBank mRNA | AK129328.1 | - | 9 - 10 | 1640 | 70 | |
| AK141959.1 | - | 9 - 10 | 1630 | 70 | ||
| AK149644.1 | - | 4 - 5 | 534 | 70 | ||
| AK159020.1 | - | 9 - 10 | 1632 | 70 | ||
| AK159754.1 | - | 9 - 10 | 1645 | 70 | ||
| BC027225.1 | - | 9 - 10 | 1457 | 3486732 | 70 | |
| L07924.1 | - | 9 - 10 | 1635 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ralgds
Genomic Map